HPLC Columns, YMC-Pack Diol-SEC
Supplier: YMC America,Inc
Certificates
About this item
Diol-SEC is a size exclusion chromatography column utilizing a dihydroxypropyl stationary phase bonded to a silica gel base and useful for molecular weight determination of proteins and sugars.
- Silica-based column with high mechanical strength
- Very low nonspecific adsorption of proteins
- More accurate quantitation of monoclonal antibodies with two products
- Provides improvement in resolution and throughput on size exclusion chromatography
Diol-120, 200, and 300 are suitable for separation or molecular weight determination of proteins with molecular weights of 10000 to several hundred thousand. Diol-60 is the most suitable for separation of peptides of oligosaccharides whose molecular weights are 10000 or less.
Specifications
- Catalog No:
- 97030-806
- 97030-804
- 97030-802
- 89216-682
- 97030-760
- 89216-686
- 97030-794
- 97030-792
- 97030-790
- 97030-832
- 97030-758
- 97030-800
- 97030-798
- 97030-754
- 97030-796
- 97030-752
- 97030-822
- 97030-830
- 97030-826
- 97030-824
- 89198-934
- 89216-698
- 89198-930
- 89216-690
- 89216-696
- 97030-772
- 97030-778
- 97030-774
- pH range:
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5
- 5 - 7.5