Cleanert® NH₂ SPE Columns, Bonna-Agela
Supplier: Agela Technologies
Certificates
About this item
Cleanert® NH₂ are silica-based, sorbent bonded with aminopropyl functional group.
- Can be used in normal or reversed phase mode
- Weak anion exchange
- Silica-based
- Sorbent bonded
Cleanert® NH₂ are silica based sorbent bonded with aminopropyl functional group. This sorbent can be used in either normal phase or reversed phase mode. It retains the analytes either by a polar adsorption (from non-polar solution) or by weak anion exchange (from aqueous solution). pKa=9.8.
Specifications
- Catalog No:
- 97018-054
- 97018-042
- 97018-046
- 97018-050
- 97018-052
- Description:
- 97018-054
- 97018-042
- 97018-046
- 97018-050
- 97018-052
- Surface area:
- 480 m2/g
- 480 m2/g
- 480 m2/g
- 480 m2/g
- 480 m2/g