ZetexPlus 220 Extreme Temperature Mitts, Double Palm, Newtex Industries
Supplier: Newtex Industries
Certificates
About this item
ZetexPlus 220 mitts are for high-temperature applications.
- Flame-resistant
- Double palm
- Operating Temperature: 815 °C (1500 °F)
- Maximum Temperature: 1095 °C (2000 °F)
Offers excellent resistance to flames, sparks, and molten splash. Non-asbestos, high-temperature replacement for high-silica and ceramic fabrics. High tensile strength. Vermiculite.
Customers Who Bought This Also Bought