N-DEX® NightHawk® Gloves, Showa
Supplier: Showa
Certificates
About this item
These nonsterile, powder-free, textured finish ambidextrous gloves protect hands from a wide array of chemical hazards, while avoiding Type I and Type IV latex allergies.
- 100% accelerator-free nitrile
- Easy to put on and remove
- Cuff prevents dirt from entering the glove
- Surface provides tactile feel and optimal grip
- Great freedom of movement for the wrist
- Gloves feature ESD and static-dissipative properties
They are suitable for light assembly tasks, handling disposables, automotive applications, and chemical industries.
Certifications: EN 374; Cat. III; AQL 1,5.
Specifications
- Catalog No:
- 80086-400
- 80086-398
- 80086-396
- 80086-394
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Anti-static:
- Yes
- Yes
- Yes
- Yes