Pierce™ Immobilised TCEP Disulfide Reducing Gel, Thermo Scientific
Catalog #: CAPI77712
Supplier: Thermo Scientific
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Tris[2-Carboxyethyl]phosphine Disulfide (TCEP), Immobilized Reducing Gel
- Size:5 mL
- Molecular Weight:286.65 g/mol
- Application:Reduces Small Peptides that Cannot Be Desalted Before Immobilization
- Supplier No.:77712
Specifications
About this item
Immobilised TCEP Disulfide Reducing Gel eliminates the need to use laborious and inefficient gel filtration methods to separate the reduced sample from the reducing agent.
- Odourless and virtually no loss of activity when stored refrigerated
- Reduces peptides and proteins over a broad range of pH, salt or detergent concentrations, and temperature conditions
- Resin slurry provides flexibility for use in spin tube, column or batch modes
- No soluble reductant removal saves time and conserves sample
- Cleaves both natural disulfide bonds and those incorporated by crosslinking
Customers Who Bought This Also Bought