About this item
Whatman™ Mini-UniPrep™ G2 filters from Cytiva can be used with either a hand compressor (single position) or a multicompressor.
- Mini-UniPrep™ vials can be loaded directly into the autosampler
The Mini-UniPrep™ hand compressor is a time reducing compressor for the Mini-UniPrep™ G2 syringeless filter, a fast and cost-effective syringeless filter that simplifies UHPLC and HPLC sample filtration. Ergonomic hand compressor allows for sample preparation with increased hand comfort. Mini-UniPrep™ all-in-one syringeless filter prepares samples three times faster than standard syringe filter method. Mini-UniPrep™ glass or polypropylene vials support analyses such as pharmaceutical quality control (pharma QC) and water monitoring.
Mini-UniPrep™ G2 multicompressor saves time and reduces hand stress by filtering multiple Mini-UniPrep™ G2 syringeless filter vials during sample preparation. Streamlines syringeless filtration by filtering up to eight filter vials simultaneously. Designed for Cytiva's Mini-UniPrep™ G2 - with integral borosilicate glass autosampler vial, plunger and attached filter membrane, and septum with cap. Glass construction prevents interference of leachables from plastic housing with HPLC/UHPLC pharmaceutical analysis.
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
GE Healthcare Environmental testing for water air and soil Brochure (CAN) GE Healthcare Environmental testing for water air and soil Brochure (US) GE Healthcare Environmental Testing for Water Brochure GE Healthcare Environmental Testing for Water Brochure (CA) GE_Healthcare_Whatman_Filtration_Products_for_Pharmaceutical_Quality_Control_(US)