Polyethylene Plug Caps, Starplex®
Supplier: Starplex
Certificates
About this item
Nonsterile polyethylene plug style caps are designed for use with 5mL and 10mL vials
All caps are bulk packaged in bags.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.