Quartz Boiling Flask, Technical Glass Products
Supplier: Technical Glass Products
Certificates
About this item
Fused Quartz consists of high-purity Silica (SiO2) with a purity level of 99.998%
Fused Quartz also offers a number of unique properties that make it ideal in a variety of applications where other materials prove to be problematic. These boiling flasks provide extremely high purity, a high softening temperature (1670°C), optical transparency over a wide range, low thermal expansion, excellent thermal shock, and resistance to a large number of elements and compounds.
Boiling Flasks manufactured from this material are invaluable in research and industry applications.
Customers Who Bought This Also Bought