To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Ethanol Assay Kit
- Size:100 assays
- Method:OD565 nm (Enzymatic)
- Sample:Serum., plasma, urine, saliva, etc.
- Processing time:30 minutes
- Cat. no.:75878-060
- Supplier No.:ECET-100
Specifications
About this item
For quantitative determination of alcohol and its metabolism.
- Linear detection range 0.0008 - 0.1 vol % (140 - 17,500 μM, or 8 -1000 ppm) in 96-well plate assay
- Can be readily automated to process thousands of samples per day
Alcoholic drinks are among the daily consumed beverages
BioAssay Systems’ EnzyChrom™ ethanol assay kit is based on alcohol dehydrogenase catalyzed oxidation of ethanol, in which the formed NADH is coupled to the formazan (MTT) chromogen. The intensity of the product color, measured at 565 nm, is proportionate to the ethanol concentration in the sample.
Studies have shown heavy alcohol consumption may lead to various forms of liver diseases and to increased mortality rates. Quantitative determination of alcohol (ethanol, C2H5OH) has applications in basic research, drug discovery, clinic studies and in the alcoholic industry. Simple, direct and automation-ready. Procedures for measuring ethanol concentration are very desirable.