FlexTech™ Y9236 Hi-Vis Cut Resistant Gloves with Nitrile Palm, Wells Lamont
Supplier: Wells Lamont
Certificates
About this item
FlexTech™ Y9236 high visibility cut resistant gloves feature a nitrile palm for improved grip.
- 15-gauge seamless knit
- Hi-vis shell
- Reinforced thumb crotch
- Knit wrist construction
- Blend of high performance and cooling fibers
- Sandy nitrile palm coating
- Outstanding fit, comfort and flexibility
FlexTech™ Y9236 gloves feature high visibility knit shell with nitrile coating that ensures workers' compliance on the job. Reinforced thumb crotch with yellow Kevlar stitching provides extra strength, while the knit wrist construction prevents dirt and debris from entering the glove. These gloves are made from a blend of high performance and cooling fibers. Sandy nitrile palm coating provides excellent grip for an outstanding fit, comfort and flexibility.
Certifications: ANSI Cut Level 4, A4. ANSI level 4 abrasion.
Ordering information: These gloves are sold by the dozen pair.
Specifications
- Catalog No:
- 75875-674
- 75875-672
- 75875-670
- 75875-668
- 75875-678
- 75875-676
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Gauge:
- 15
- 15
- 15
- 15
- 15
- 15