Specifications
- Description:Aspartate Assay Kit
- Size:100 assays
- Method:OD570 nm, or FL530/585 nm
- Sample:Plasma, serum, tissue, culture media etc.
- Processing time:60 minutes
- Cat. no.:75878-046
- Supplier No.:EASP-100
Specifications
About this item
For quantitative determination of aspartate and evaluation of drug effects on aspartate metabolism.
- Linear detection range 2 to 400 μM aspartate for colorimetric assays and 1 to 50 μM for fluorimetric assays
Aspartate, a nonessential amino acid, is a precursor to several other amino acids and is an excitatory neurotransmitter
BioAssay Systems’ Aspartate Assay Kit provides a simple, direct and automation-ready procedure for measuring aspartate concentration. Aspartate is converted into pyruvate which is then oxidized with the conversion of the dye into a colored and fluorescent form. The color intensity of the oxidized dye at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the aspartate concentration in the sample.
Aspartate is involved in the urea cycle and gluconeogenesis.