Specifications
- Description:Starch Assay Kit
- Size:100 assays
- Method:OD570 nm, or FL530/585 nm
- Sample:Biological, agriculture, food etc.
- Processing time:30 minutes
- Cat. no.:75878-028
- Supplier No.:E2ST-100
Specifications
About this item
For quantitative determination of starch and evaluation of drug effects on starch metabolism.
- Linear detection range 2 - 200 μg/mL starch for colorimetric assays and 0.2 - 20 μg/mL for fluorimetric assays
Starch, chemical formula (C6H10O5)n, is a polysaccharide carbohydrate consisting of a large number of glucose units joined together by glycosidic bonds
BioAssay Systems’ starch uses a single Working Reagent that combines the enzymatic break down of starch and the detection of glucose in one step. The color intensity of the reaction product at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the starch concentration in the sample. This simple convenient assay is carried out at room temperature and takes only 30 min.
All plant seeds and tubers contain starch present in the form of amylose and amylopectin. Starch is the most consumed polysaccharide in the human diet. Some starches are digested very quickly, and cause a rapid and large rise in blood sugar. Others are digested more slowly, and some starch, called resistant starch, is not digested in the small intestine at all, and thus causes little or no blood sugar rise. Simple, direct and automation-ready. Procedures for measuring starch concentrations find wide applications in research and drug discovery.