ContactUsLinkComponent
QuantiChrom™ Heme Assay Kit, BioAssay Systems
QuantiChrom™ Heme Assay Kit, BioAssay Systems
Catalog #: 75877-998
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
QuantiChrom* Heme Assay Kit 250 tests
QuantiChrom™ Heme Assay Kit, BioAssay Systems
Catalog #: 75877-998
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Heme Assay Kit
  • Size:
    250 assays
  • Method:
    OD400 nm
  • Sample:
    Blood, serum, plasma, urine etc.
  • Cat. no.:
    75877-998
  • Supplier No.:
    DIHM-250
Specifications
About this item
For quantitative determination of heme and evaluation of drug effects on heme metabolism.
  • Safe
  • Reagents are non-toxic
  • Linear detection range 0.6 - 125 μM heme
  • The "mix-and-read" 5 minute procedure
  • Can be readily automated to process tens of thousands of samples per day
  • Assays can be executed in microplate or cuvet
Heme is one important member of the porphyrin family. It is synthesized in both mitochondria and cytoplasm, and is a key prosthetic group for various essential proteins such as hemoglobin, cytochromes, catalases and peroxidases. Heme determination is widely practiced by researchers of various blood diseases. Simple, direct and automation-ready. Procedures for measuring heme concentration are becoming popular in Research and Drug Discovery. BioAssay Systems’ QuantiChrom™ Heme Assay Kit is based on an improved aqueous alkaline solution method, in which the heme is converted into a uniform colored form. The intensity of color, measured at 400 nm, is directly proportional to the heme concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples and exhibits high sensitivity.
Customers Who Bought This Also Bought