To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Acetylcholinesterase Assay Kit
- Size:100 assays
- Method:OD412nm
- Sample:Blood, serum, plasma etc.
- Processing time:10 minutes
- Cat. no.:75877-948
- Supplier No.:DACE-100
Specifications
About this item
For quantitative determination of acetylcholinesterase activity.
- Detection range: 10 to 600 U/L AChE activity in 96-well plate assay
- Can be readily automated as a high-throughput assay for thousands of samples per day
Acetylcholinesterase (EC 3.1.1.7, AChE), also known as RBC cholinesterase, is found primarily in the blood and neural synapses
BioAssay Systems’ QuantiChrom™ Acetylcholinesterase Assay is based on an improved Ellman method in which thiocholine produced by the action of acetylcholinesterase forms a yellow color with 5,5’-dithiobis(2-nitrobenzoic acid). The intensity of the product color, measured at 412 nm, is proportional to the enzyme activity in the sample.
AChE catalyzes the hydrolysis of the neurotransmitter acetylcholine into choline and acetic acid, a reaction necessary to allow a cholinergic neuron to return to its resting state after activation. Studies show serum cholinesterase activity may relate to insecticide exposure or to one of a number of variant genotypes. As a result, cholinesterase levels of cells and plasma are used as a guide in establishing safety precautions relative to exposure and contact, as well as a guide in determining the need for workers to be removed from areas of contact with the organic phosphate insecticides.Simple, direct and automation-ready procedures for measuring AChE activity are highly desirable.