VWR® Economy Timer
Catalog #: 89140-544
Supplier: VWR International
Specifications
- Description:Economy timer
- Cat. no.:89140-544
Specifications
About this item
Timer can be programmed in countdown mode or countup mode up to 99 minutes, 59 seconds.
- Count up and down simultaneously
- Big digit display
Delivery: Supplied with loud buzzer, magnet, clip, and stand.
Customers Who Bought This Also Bought