About this item
3D printing made affordable! A lightweight and eco-friendly model yet is perfect for starter lab.
- Single nozzle print head
- Easy calibration
- Eco friendly materials
- Compact design
- FFF (Fused Filament Fabrication) technology
- PLA filament material
- Multi language
Traditional 3-to-4 knob calibrations on the beds of 3D printers are complicated, but with the da Vinci Jr. 1.0, the print bed is already completely leveled and it only requires a Z Offset calibration. With the Z Offset calibration, you can adjust the distances between the extruder and the print bed, making sure that each print comes out perfectly.
The DaVinci Jr. features a fluid, press-and-release extruder which makes for an easy extruder replacement or cleaning experience. Simply open up your 3D printer, press the release button, and the replacement or cleaning process can be done within few seconds.
The DaVinci Jr.’s PLA filament spools are a non-toxic, biodegradable plastic that is recyclable and safe for the environment! With its compact design, the DaVinci Jr. only consumes 75 watts of power. This eco-friendly 3D printer makes sure that nothing goes to waste.
With its completely enclosed designed and non-heated print tray, the DaVinci Jr. 1.0 is built with your safety in mind. Kids, beginners, and Makers can all enjoy countless hours of 3D printing. The EZ-Offset Calibration makes it easy to get perfect prints every time, while the auto-feeding filament system and easy-to-install filament spools save you time on getting your first project started.
Uses only XYZ Brand DaVinci jr filament specified below.
Specifications
- Catalog No:
- 470226-906
- Build volume:
- Computer requirements:
- Connections:
- Display:
- Supported OS:
- Ø:
- Dimensions:
- Shipping dimensions: