VeriSpec™ Elan DRC Plus Tuning Solution II, Ten Components, RICCA Chemical Company
Supplier: Ricca Chemical
Certificates
About this item
Certified reference material (CRM) for ICP-MS
10 ppb: Ba; 1 ppb: Be, Ce, Co, In, Fe, Pb, Mg, Th, U
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.