VeriSpec® Sodium (Na) Standard for AAS 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
Certificates
About this item
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.