SP Wilmad-LabGlass Complete Vacuum Traps with 1/4" or 1/2" Hose Connections, SP Industries
Supplier: SP Industries
Certificates
About this item
Use with a wide variety of Dewar Flasks.
- Borosilicate glass
These single-piece vacuum traps are designed with two vertical hose connectors which allows for use with a wide variety of Dewar Flasks.
Customers Who Bought This Also Bought