Uvex Carbonvision™ Safety Goggles, Honeywell Safety
Supplier: Honeywell Safety Products
Certificates
About this item
These safety goggles with Dura-streme® dual anti-fog/anti-scratch coating are designed with flexible soft frame which adapts perfectly to the face to ensure a perfect fit and seal around the eyes for high-performance protection and all-day comfort.
- Soft, flexible body which is permanently fixed to the frame, making it easy to clean and maintain
- Protects from impact, dust/airborne particles as well as chemical splash hazards
- Convenient clip makes it easy to remove headband before cleaning
Low profile and its compact design is ideal for wearing with disposable respirators and protective caps. High-quality headband is available in neoprene or fabric and offers a comfortable fit and long-lasting durability. Available in Clear and Gray lens tints.
Certifications: Meets the ANSI Z87.1-2010 standard. Certified to the requirements of the CSA Z94.3 standard.
Customers Who Bought This Also Bought