LaboPlast® and SteriPlast® PowderDispo, Burkle
Supplier: BURKLE INC
Certificates
About this item
The PowderDispo is a disposable sampler for cross-section samples of powdery or slightly cohesive substances. A complete all-layer samples can be taken from bags or smaller containers with the lance.
- Cleanroom manufactured
- Optionally sterilised by gamma rays
- Cross contamination is completely eliminated
Disposable sampler for the highest standards in sampling unlike conventional reusable samplers, disposable samplers do not need to be cleaned after use.
Packaging: 20 pieces per pack.
Customers Who Bought This Also Bought