Order Entry
Canada
ContactUsLinkComponent
Glycine ≥99%, Molecular Biology Grade, Calbiochem®
Glycine ≥99%, Molecular Biology Grade, Calbiochem®
Catalog # CA80502-614
Supplier:  MilliporeSigma
CAS Number:  
undefined
Glycine ≥99%, Molecular Biology Grade, Calbiochem®
Catalog # CA80502-614
Supplier:  MilliporeSigma
Supplier Number:  357002-1KG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 kg
  • Cat. no.:
    CA80502-614
  • Supplier No.:
    357002-1KG

Specifications

Specification Test Results

  • Form:
    White solid
  • Purity:
    ≥99% (by titration)
  • Contaminants:
    DNases, proteases, RNases: none detected.
  • Toxicity:
    Standard Handling
  • Storage:
    +15 °C - +30 °C
  • Solubility:
    H2O (200 mg/ml)
  • Stability:
    Following reconstitution, store at room temperature (20°C). Stock solutions are stable for up to 6 months at room temperature.

Specification Test Results

About this item

 :
 :75.07 g/mol
Storage Temperature:Ambient
CAS Number:56-40-6
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131