N-Acetylneuraminic acid (synthetic) ≥98% (by HPLC), Calbiochem®
Catalog #: CA80055-610
Supplier: MilliporeSigma
CAS Number: 131-48-6
Specifications
- Packaging:Plastic ampoule
- Size:1 g
- Cat. no.:CA80055-610
- Supplier No.:110138-1GM
Specifications
Specification Test Results
- Form:White solid
- Hygroscopic:Hygroscopic
- Purity:≥98% by HPLC
- Toxicity:Standard Handling
- Storage:−20 °C, Protect from Light
- Stability:Following reconstitution, aliquot and freeze (-20 °C). Stock solutions are stable for up to 6 months at -20 °C
- Calbiochem®, N-Acetylneuraminic acid (synthetic) ≥98% (by HPLC):
Specification Test Results
About this item
Synonyms: Neu5Ac,(4S,5R,6R,7S,8R)-5-Acetamido-4,6,7,8,9-pentahydroxy-2-oxononanoic acid,NANA,Sialic acid,5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-nonulopyranosonic acid
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 309.27 g/mol
Formula:C₁₁H₁₉NO₉
MW:309.27 g/mol
MDL Number:MFCD00006620
CAS Number:131-48-6
Customers Who Bought This Also Bought