Order Entry
ContactUsLinkComponent
DRG® Candida albicans IgG ELISA, DRG International, Inc.
DRG® Candida albicans IgG ELISA, DRG International, Inc.
Catalog #: 75871-262
Supplier:  DRG International
CAS Number:  
CANDIDA ALBICANS IGG ELISA
DRG® Candida albicans IgG ELISA, DRG International, Inc.
Catalog #: 75871-262
Supplier:  DRG International
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Candida albicans IgG ELISA
  • Sample type:
    Serum
  • Application:
    Infectious Diseases
  • Regulatory status:
    FDA Regulated; CE Approved
  • Cat. no.:
    75871-262
  • Supplier No.:
    EIA-3458
  • No. of Tests:
    96 Wells

Specifications

About this item

An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Candida albicans in serum.

  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests

Species of Candida are non-mycelia-producing non-ascospore-forming yeast-like fungi which appear as small (4 to 6 µm), oval, thin-walled gram-positive budding cells. The most important representative, Candida albicans, is a facultative pathogen for man.C. albicansis ubiquitous and commonly found as transient flora on normal mucous membranes. Although not pathogenic in healthy humans the fungus may be opportunistic in those suffering from a variety of disorders, and in those treated intensively with broad-spectrum antibiotics or immunosuppressive measures. Candidiasis is caused about 90% of the time byC. albicans. It is an acute or subacute infection in which the fungus may produce lesions in the mouth (thrush, oral candidiasis), vagina (vulvovaginal candidiasis), skin and nails (intertriginous candidiasis), bronchi or lungs (bronchopulmonary candidiasis) and, occasionally, a septicemia, endocarditis or meningitis. In immunosuppressant patients with cellular immunodeficiency, e.g., AIDS patients,C. albicansmay lead to severe necrosis of infected tissues. The DRG Candida albicans IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) Microtiter wells as a solid phase are coated with Candida albicans antigen. Diluted patient specimens and ready-for-use controls are pipetted into these wells. During incubation Candida albicans-specific antibodies of positive specimens and controls are bound to the immobilized antigens. After a washing step to remove unbound sample and control material horseradish peroxidase conjugated anti-human IgG antibodies are dispensed into the wells. During a second incubation this anti‑IgG conjugate binds specifically to IgG antibodies resulting in the formation of enzyme-linked immune complexes. After a second washing step to remove unbound conjugate the immune complexes formed (in case of positive results) are detected by incubation with TMB substrate and development of a blue color. The blue color turns into yellow by stopping the enzymatic indicator reaction with sulfuric acid. The intensity of this color is directly proportional to the amount of Candida albicans-specific IgG antibody in the patient specimen. Absorbance at 450 nm is read using an ELISA microtiter plate reader.

Customers Who Bought This Also Bought