Order Entry
ContactUsLinkComponent
DRG® DHEA-S ELISA, DRG International
DRG® DHEA-S ELISA, DRG International
Catalog #: 75871-124
Supplier:  DRG International
CAS Number:  
DHEA-S ELISA
DRG® DHEA-S ELISA, DRG International
Catalog #: 75871-124
Supplier:  DRG International
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type:
    Competitive
  • Description:
    DHEA-S ELISA
  • Sample type:
    Serum
  • Detection range:
    0 – 10 µg/ml
  • Application:
    Fertility/Pregnancy
  • Regulatory status:
    FDA Regulated; CE Approved
  • Cat. no.:
    75871-124
  • Supplier No.:
    EIA-1562
  • No. of Tests:
    96 Wells

Specifications

About this item

An enzyme immunoassay for the quantitative measurement of DHEA‑S in serum and plasma.

  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests

Dehydroepiandrosterone (5-Androstene-3β-0L-17-one, Androstenolone, Dehydroisoandrosterone, Transdehydroandrosterone, DHEA) is a steroid hormone present in blood mostly in its sulfate form (DHEA S)

The DRG DHEA‑S ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal antibody directed towards an antigenic site of the DHEA‑S molecule. Endogenous DHEA‑S of a patient sample competes with a DHEA‑S-horseradish peroxidase conjugate for binding to the coated antibody. After incubation the unbound conjugate is washed off. The amount of bound peroxidase conjugate is inversely proportional to the concentration of DHEA‑S in the sample. After addition of the substrate solution, the intensity of colour developed is inversely proportional to the concentration of DHEA‑S in the patient sample.

DHEA-S is a more specific product of the adrenals and measurements of this steroid are widely used in clinical practice. The clinical importance of plasma assays of DHEA-S is associated with the diagnosis of adrenal hyperplasia and differential diagnosis of hirsutism.

Customers Who Bought This Also Bought