Difco™ m FC Broth Base, BD Diagnostics
Supplier: Avantor
Certificates
About this item
Use with Rosolic Acid For detection and enumeration of fecal coliforms by the membrane filter technique at elevated temperatures.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.