Order Entry
Canada
ContactUsLinkComponent
Taq-&GO™ Ready To Use PCR Mix, MP Biomedicals, LLC
Taq-&GO™ Ready To Use PCR Mix, MP Biomedicals, LLC
Catalog # CA76046-310
Supplier:  MP Biomedicals
CAS Number:  
Taq-&GO™ Ready To Use PCR Mix, MP Biomedicals, LLC
Catalog # CA76046-310
Supplier:  MP Biomedicals
Supplier Number:  11EPTAG110
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Description:
    Ready To Use PCR Mix Molecular Biology Grade
  • Environmentally Preferable:
  • Storage conditions:
    −20 °C
  • Concentration:
    5X
  • Cat. no.:
    CA76046-310
  • Volume:
    10× 1 ml

Specifications

About this item

Taq-&GO™ is a ready-to-use, optimized reaction mix for PCR and is provided 5X concentrated.

  • Molecular Biology Grade
  • Optimized for highly efficient PCR
  • Significant savings in preparation time
  • Reduced pipetting
  • Reduced risks of errors and contamination
  • Stable at 4 °C

All MP Biomedical thermostable DNA polymerases are available in ready-to-use &GO™ PCR mastermix formats. &GO™ PCR Mastermixes contain all the components for PCR with the exception of template and primers. This not only saves valuable time in the laboratory,but also reduces the risk of pipetting and reagent handling errors. Each &GO™ mix is provided 5Xconcentrated and can be used to perform 20 μL or 50 μL reactions. To prepare the reaction, simply add 10 μL of &GO™ PCR mastermix 5XC to template and primers diluted to 40 μL. For smaller reactions, use 4 μL of mastermix in a 20 μL reaction.

Components: Taq DNA Polymerase, dNTPs, MgCl2, and buffer