About this item
The UVP PhotoDoc-It™ systems are designed for fluorescence imaging. The stand-alone systems include a transilluminator, filter, camera, printer and darkroom; no computer is required.
- DigiCam color camera with 16 megapixel resolution
- Lightweight, compact hood provides a compact darkroom environment, hood now includes a new UV blocking viewer window
- Benchtop UV Transilluminator delivers back-lit UV; select from several benchtop models
- Color printer generates instant 4×6" low cost, laboratory quality color prints
- Ethidium bromide filter for fluorescent stained gels included
- Combination lock secures the camera to the darkroom
Illuminate stained gels with the UV fluorescence of the compact UV transilluminator.
The Ethidium Bromide emission filter, positioned inside the hood, is easily interchangeable for other filter types. Capture brilliant color images with the high resolution, megapixel digital color camera. View images through the viewer window. Save images to the memory card (4GM minimum) or instantly print images on the laboratory quality photo printer included with the system. The camera can be connected to a PC with a USB cable (not included) for transferring images.
The DigiCam is a color digital camera with 16 megapixels and a 4GB (minimum) SD memory card. Power cord included (batteries not required).
The darkroom fits on top of the transilluminator and has a UV blocking viewer window for safe viewing of gels. The darkroom is lightweight and compact with handles for easy transportation of the hood.
The system is designed for use with UVP Benchtop UV Transilluminators. Select from 302nm UV or 2UV models with 302nm and 365nm. Filter sizes are 20x20cm or 21x26cm. M-20V and M-26V feature variable intensity settings.
Color printer features 300x300 resolution, 4x6" prints, and USB connectivity. 108 prints included. Combination lock includes a 3 digit resettable combination and 2 ft. cable.