About this item
Used primarily for lysis of gram positive and gram negative bacteria, fungal tissue and spores, yeasts, molds, filamentous algaes and biofilms.
- Impact-resistant tube
- Yttria-Stabilized Zirconium oxide beads have a high density and extremely high hardness
- Ideal for cracking very tough cell walls
- Smooth surface minimizes shearing
Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then prepared and dispensed under a rigorous set of proprietary conditions that offer customers a premium product that can be immediately used with confidence. Matrices are available separately for use with your own unique buffers, and are also available as components of the complete purification kits.
Application: Extractions of nucleic acids, proteins, metabolites and other small molecules from organisms with very tough cell walls. Lysis of gram positive and gram negative bacteria, fungal tissue and spor es, yeasts, molds, filamentous algae and biofilms. RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA76045-740
- CA76045-738
- CA76045-758
- CA76046-086
- CA76046-084
- CA76046-082
- Storage and handling:
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Store at Room Temperature (15...30 °C)
- Molecule enriched:
- DNA, RNA, Proteins, Other small molecules
- DNA, RNA, Proteins, Other small molecules
- DNA, RNA, Proteins, Other small molecules
- DNA, RNA, Proteins, Other small molecules
- DNA, RNA, Proteins, Other small molecules
- DNA, RNA, Proteins, Other small molecules
- Materials required (not supplied):
- FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- Material of construction:
- 0.5 mm diameter Yttria-Stabilized Zirconium Oxide beads
- 0.5 mm diameter Yttria-Stabilized Zirconium Oxide beads
- 0.5 mm diameter Yttria-Stabilized Zirconium Oxide beads
- 0.5 mm diameter Yttria-Stabilized Zirconium Oxide beads
- 0.5 mm diameter Yttria-Stabilized Zirconium Oxide beads
- 0.5 mm diameter Yttria-Stabilized Zirconium Oxide beads
- Instrument Compatibility:
- FastPrep-24, FastPrep-96, FastPrep-24 5G
- FastPrep-24, FastPrep-96, FastPrep-24 5G
- FastPrep-24, FastPrep-96, FastPrep-24 5G
- FastPrep-24, FastPrep-96, SuperFastPrep-2, FastPrep-24 5G
- FastPrep-96
- FastPrep-96
- Adapter compatibility:
- TeenPrep (12×15 ml), CoolTeenPrep (6×15 ml), TeenFlex (24×15 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- TeenPrep (12×15 ml), CoolTeenPrep (6×15 ml), TeenFlex (24×15 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- TeenPrep (12×15 ml), CoolTeenPrep (6×15 ml), TeenFlex (24×15 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- 96-Well Adapter
- 96-Well Adapter
- Application areas:
- Sample Preparation, Sample Lysis, Nucleic Acid Isolation & Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation
- Sample Preparation, Sample Lysis, Nucleic Acid Isolation & Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation
- Sample Preparation, Sample Lysis, Nucleic Acid Isolation & Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation
- Sample Preparation, Sample Lysis, Nucleic Acid Isolation & Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation
- Sample Preparation, Sample Lysis, Nucleic Acid Isolation & Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation
- Sample Preparation, Sample Lysis, Nucleic Acid Isolation & Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation