Human Chromosome Genome Study Kit
Catalog #: 470100-572
Supplier: KELLER BROS. & MILLER, INC.
Specifications
- Length:76 cm
- Width:51 cm
- Description:Human Chromosome Map Notebook Charts
- Cat. no.:470100-572
- Finish:Unlaminated
- Supplier No.:4517602
Specifications
About this item
Teaching tool for evolution and speciation, and DNA and protein synthesis.
- Identify certain gene loci
- Understand how gene mutations can cause diseases
Use this human genome study kit to teach students how to identify certain gene loci and to understand how gene mutations can cause diseases like cystic fibrosis and sickle cell anemia. It's also a perfect teaching tool for lessons on evolution and speciation, and DNA and protein synthesis. We've included a color wallchart of 22 autosomes and the X and Y sex chromosomes, with labeled genes that code for proteins such as hormones and enzymes.
Customers Who Bought This Also Bought