Trajan, PEEK Fingertight Fittings, Trajan Scientific and Medical
Catalog #: 10799-764
Supplier: Trajan Scientific and Medical
Specifications
- Description:PEEK Fingertight Fitting, Biocompatible
- Material:PEEK
- Cat. no.:10799-764
- Fitting Size:¹⁄₃₂" - 6 - 40 UNF
- Supplier No.:222017
Specifications
About this item
PEEK Fingertight fittings offer absolute zero to low dead volume connections and are ideal for bioanalytical applications.
- Unique design provides Absolute Zero or low dead volume connections
- Biocompatible material
Customers Who Bought This Also Bought