Pans, Stainless Steel, Carver
Catalog #: 53880-010
Supplier: Carver
Specifications
- Description:Stainless Steel Pan
- Cat. no.:53880-010
- Supplier No.:237005
Specifications
About this item
Stainless steel fluid pan for use with all Test Cylinder Outfits.
When presses are used for fluid extraction, filter pads sandwich the sample. Stainless steel pans capture the released fluid.
Customers Who Bought This Also Bought