Orders LinkContactUsLinkComponent
Glass Glycol Bottle, Dickson
Glass Glycol Bottle, Dickson
5 ml, Glycol bottle, Glass
Catalog # 89230-222
Glass Glycol Bottle, Dickson
Catalog # 89230-222
Supplier:  
Glass Glycol Bottle, Dickson
5 ml, Glycol bottle, Glass
Discontinued
Product Details & Documents
Glycol Bottles are a great way to stabilize your temperature readings when using a probe.
  • Can be used to mimic the substances you are measuring
  • Compatible with any K-TC beadwire probes
This is to stabilize a temperature reading in a refrigerator that is opened and closed often. The air temperature in the refrigerator (or any space) may change faster than the temperature of the material you are monitoring and the glycol bottles may better represent the true temperature of the monitored materials.
Specifications
  • Capacity:
    5 ml
  • Description:
    Glycol bottle
  • Material:
    Glass