Diamond-Plate with GRITSHIELD™ UltraSoft Anti-Fatigue Mat, Wearwell®
Supplier: Wearwell
Certificates
About this item
GRITSHIELD™ surface delivers 40% more traction.
- Chemical and abrasion resistant
- 2X the resilience and 3X the life of look-alikes
- Maximized standing area
- Unique pre-bond bevel and lay-flat design prevent trip hazards
The unique Nitricell® sponge base delivers twice the resilience and three times the life expectancy of the simple PVC sponge used in the matting industry. The pre-beveled edge assures a perfect lay-flat quality. The sponge is extended within 1.5" of the mat's surface for 20% more standing area than others in the market. Ideally suited for use in dry environments.
Certifications: Meets ASTM F1677. Dry: COF = 1.10/Wet: COF = 1.0. Meets ASTM D575. Deflection @35 psi = 0.50.
Customers Who Bought This Also Bought