Accessories for Overhead Stirrers VOS Range, IKA
Catalog #: 33994-096
Supplier: IKA WORKS, INC. MX
Specifications
- Description:FK 1 Flexible Coupling
- Cat. no.:33994-096
- Supplier No.:2336000
- Details:For Applications Using Glass Stirring Rods (Diameter 6-10 mm), Max. Torque 10 Ncm