Microchem® by AlphaTec™ 68-CFR Flame Retardant Coveralls with Serged and Taped Seams, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
MICROCHEM® by AlphaTec™ CFR is a flame retardant and antistatic fabric offering protection from particulates and light liquid spray or splash without compromising worker protection in the event of a flash fire.
- Flame retardant treated fabric with PVC barrier film offering wearers protection from liquid chemicals
- Versatile in most applications where there is the need for protection from chemical spray without compromising wearer protection in the event of a flash fire
- Optimized body fit improves wearer comfort and safety
- Highly visible bright red color to improve worker safety
Must be worn over thermal protective garments, such as NOMEX®, and never to be worn next to the skin. These disposable latex coveralls feature an elasticated hood, wrists, and ankles with double zipper closure. Typically used in oil and petrochemical industries, petroleum distribution and processing, and utilities.
Certifications: Tested according to EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN 14605 Type 3, EN 14605 Type 4, EN ISO 13982-1 Type 5, EN ISO 14116 INDEX 1/0/0, ANSI/ISEA 101.
Specifications
- Catalog No:
- 10836-460
- 10836-458
- 10836-462
- 10836-280
- 10836-278
- 10836-456
- 10836-276
- 10836-274
- Fabric:
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Seam type:
- Taped
- Taped
- Taped
- Taped
- Taped
- Taped
- Taped
- Taped
- Garment application:
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory