Microchem® by AlphaTec™, 68-5000 Chemical Protection Coveralls with Hood and Ultrasonically Welded and Taped Seams, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
MICROCHEM® by AlphaTec™ 5000 reaches new levels in chemical protection and has been engineered to protect. The highly visible multi-layer fabric is strong, durable, and suitable for workers in extremely hazardous areas, including HAZMAT response teams.
- Barrier to numerous organic and inorganic chemicals and biological hazards
- Multi-layer material, which is lightweight yet strong and durable
- Innovative design features include liquid-tight dual zip closure without the need for additional taping
- Bright orange color for improved worker safety
Breakthrough time against 14 of 15 chemicals listed in EN ISO 6529 >480 minutes
Typical areas of application include industrial and tank cleaning, sewage purification installations, chemicals, oil and petrochemicals, mining, agriculture, HAZMAT emergency response (i.e. level B), first response, and fire service.
Coveralls feature a 2-piece hood, double zip closure, and double cuffs with a knitted inner cuff.
Certifications: Tested according to EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B, EN ISO 13982-1 Type 5-B, ANSI/ISEA 101.
Specifications
- Catalog No:
- 10836-452
- 10836-448
- 10836-454
- 10836-250
- 10836-444
- 10836-254
- 10836-252
- Fabric:
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Seam type:
- Taped
- Taped
- Taped
- Taped
- Taped
- Taped
- Taped
- Garment application:
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory