Microchem® by AlphaTec™ 68-3000 Chemical Protection Coveralls, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
MICROCHEM® by AlphaTec™ 3000 features one of the lightest and most comfortable chemical protective materials on the market today
- Multi-layer barrier fabric effective against numerous chemicals
- Bright yellow for improved worker safety
- Typical coverall features include dual zip systems and double cuffs
- Antistatic tested according to EN1149-5 and AATCC76
Typical areas of application include chemicals, oil and petrochemicals, pharmaceutical, food industry (caustic clean downs), sewage purification installations, industrial and tank cleaning, and mining.
This durable multilayer fabric provides an extremely effective barrier against both inorganic chemicals and biological hazards.
Certifications: Tested according to EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B, EN ISO 13982-1 Type 5-B, ANSI/ISEA 101.
Specifications
- Catalog No:
- 10836-434
- 10836-222
- 10836-432
- 10836-220
- 10836-224
- 10836-430
- 10836-218
- 10836-428
- Fabric:
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Seam type:
- Ultrasonically welded
- Ultrasonically welded
- Ultrasonically welded
- Ultrasonically welded
- Ultrasonically welded
- Ultrasonically welded
- Ultrasonically welded
- Ultrasonically welded
- Garment application:
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory