Microchem® by AlphaTec™ 68-2300 Lightweight Chemical Protection Coveralls with Bound Seams, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
MICROCHEM® by AlphaTec™ 2300 provides an excellent barrier to harmful chemicals and blood borne pathogens, while being lightweight and relatively strong and durable.
- Lightweight yet strong and durable
- Typical coverall features include respirator fit hood and zip flap with self-adhesive tape closure
- Finger loops
- Antistatic tested according to EN1149-5 and AATCC76
These disposable latex coveralls provide a barrier to numerous inorganic liquid chemicals including acids and bases. They achieve the highest classifications for protection from biological agents in accordance with EN 14126:2003 and ASTM F1671 for penetration of blood, bodily fluids, and blood-borne pathogens. Coveralls feature an elasticated hood, waist, wrists, and ankles, along with a 2-way front zipper with resealable storm flap. These garments can be used in environmental clean-up, sewage purification installations, and composites manufacturing.
Certifications: Tested according to EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, ANSI/ISEA 101.
Specifications
- Catalog No:
- 10836-412
- 10836-410
- 10836-208
- 10836-186
- 10836-206
- 10836-188
- 10836-200
- 10836-198
- 10836-204
- 10836-202
- 10836-192
- 10836-190
- 10836-196
- 10836-194
- 10836-416
- 10836-414
- Fabric:
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Chemical barrier laminate
- Seam type:
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Garment application:
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory