Microchem® by AlphaTec™ 68-1500 Industrial Coveralls with Serged Seams, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
These MICROCHEM® by AlphaTec™ 1500 coveralls have been designed for workers involved in the stripping, clean-up, or handling of asbestos, as well as general maintenance, construction, and contract cleaning.
- Air and water vapor permeable to help reduce the risk of heat stress
- Silicone free which is critical in spray painting applications
- Filter size of particles >3 microns tested with JSTIIF particle penetration test
- Optimized body fit improves wearer comfort and safety
Serged seams combine strength with particle barrier capabilities. Garments feature elasticated hood, wrists, and ankles, and a 2-way front zipper with storm flap.
Certifications: Tested according to ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6.
Specifications
- Catalog No:
- 10836-122
- 10836-120
- 10836-090
- 10836-138
- 10836-136
- 10836-094
- 10836-092
- 10836-140
- 10836-130
- 10836-128
- 10836-134
- 10836-132
- 10836-098
- 10836-096
- 10836-100
- 10836-114
- 10836-112
- 10836-118
- 10836-116
- 10836-106
- 10836-104
- 10836-110
- 10836-108
- Fabric:
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- Seam type:
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Serged
- Garment application:
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory