Square Blade Impellers, Caframo
Supplier: Caframo
Certificates
About this item
These tangential flow impellers are effective for low speed, high viscosity mixing
The square blade is welded to an 8 mm (5/16") diameter shaft, all electropolished 316L stainless steel. For stirring small volumes of viscous product.
Customers Who Bought This Also Bought