SP Wilmad-LabGlass Volatile Oil Distilling Apparatus for Oils Lighter than Water, SP Industries
Supplier: SP Industries
Certificates
About this item
Designed for oils lighter than water, this distilling apparatus is made of borosilicate glass and consists of a Clevenger-type trap with a Standard Taper 24/40 outer joint on top and same size inner joint the sidearm
A 250mL long neck round bottom flask (LG-7221) and a cold finger condenser with a Standard Taper 24/40 inner joint complete the assembly.
Customers Who Bought This Also Bought