Specifications
- Description:Ward's® Halides: Family Characteristics Lab Activity
- No. of Lab Groups:15
- Cat. no.:470023-824
- Includes:Teacher's Guide with SDS, Student Copy Master
- Supplier No.:WARD470023-824
Specifications
About this item
Observe the Chemical Properties of Element Families
- Students explore the periodic table
- Can be completed in 40-50 minutes
Through the process of combining oxygen and halide family compounds with metal nitrates and copper sulfate, students will observe and compare the chemical reactivities of various elements.