SP Wilmad-LabGlass Volatile Oil Distilling Apparatus for Oils Heavier than Water, SP Industries
Supplier: SP Industries
Certificates
About this item
Designed to be used with oils heavier than water, this borosilicate glass distilling apparatus consists of a Clevenger-type oil trap with a Standard Taper 24/40 top outer joint and an inner joint on the sidearm.
Included are a round bottom long neck flask (LG-7221) and a cold finger condenser with a Standard Taper 24/40 inner joint.
Customers Who Bought This Also Bought