n-Hexane ≥97.0% (by GC), CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
Synonyms: Hexyl hydride,Hexanen
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MW:86.18 g/mol
Boiling Pt:66 - 69 °C(1.013 hPa)
MDL Number:MFCD00009520
:n-Hexane
Melting Pt:-95 °C
Density:0.660-0.668 g/cm³ (20 °C) (lit.)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97,0
Flash Pt:7.4 %(V)
:Boiling Pt:
:Boiling Pt: 66 - 69 °C(1.013 hPa)
:Melting Pt:
:Melting Pt: -95 °C
:Flash Pt:
:Flash Pt: 7.4 %(V)
:M.W. 86.18 g/mol
EINECS:203-777-6
:66 - 69 °C(1.013 hPa)
UN:1208
:3
:II
:7.4 %(V)
:GHS07
:GHS08
:GHS02
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (GC):Min. 97.00 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.010 %
- Free acid (as CH₃COOH):Max. 0.001 %
- Absorbance at 250 nm:Max. 0.01
- Absorbance at 225 nm:Max. 0.1
- Absorbance at 220 nm:Max. 0.7
- Absorbance at 210 nm:Max. 0.3
- Absorbance (JP)(Val.Ref):complying
- IR spectrum:complying
- Peroxides JP:complying
- Vapor Pressure:~132 mmHg ( 20 °C)
- Refractive Index:n20/D 1.375 (lit.)
- Vapor Density:~3 (vs air)
- Riedel-de Haën™, n-Hexane ≥97.0% (by GC), CHROMASOLV™ for HPLC:
Specification Test Results
Related Information
Customers Who Bought This Also Bought