About this item
BioPulverizer™ System I contains 10x2mL tubes of each of our most popular Lysing Matrix types (A, B, C, D and E).
- Great starter pack for new FastPrep instrument owners
- Optimize homogenization conditions
- Contains 10 of each matrix: A, B, C, D and E
- Impact-resistant 2 mL tubes
Matrix A, contains garnet matrix and one 1/4 inch ceramic sphere. Matrix B, contains 0.1 mm silica spheres. Matrix C contains 1 mm silica spheres. Matrix D, contains 1.4 mm ceramic spheres. Matrix E, Each impact-resistant 2 mL tube contains 1.4 ceramic spheres, 0.1 mm silica spheres, and one 4 mm glass bead.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA76046-186
- IC116750200
- Tube compatibility:
- 2 ml
- 2 ml
- Matrix type:
- A, B, C, D
- A, B, C, D
- Material of construction:
- Matrix A, contains garnet matrix and one 1/4 inch ceramic sphere. Matrix B, contains 0.1 mm silica spheres. Matrix C contains 1 mm silica spheres. Matrix D, contains 1.4 mm ceramic spheres. Matrix E, Each impact-resistant 2 mL tube contains 1.4 ceramic spheres, 0.1 mm silica spheres, and one 4 mm glass bead.
- Matrix A, contains garnet matrix and one 1/4 inch ceramic sphere. Matrix B, contains 0.1 mm silica spheres. Matrix C contains 1 mm silica spheres. Matrix D, contains 1.4 mm ceramic spheres. Matrix E, Each impact-resistant 2 mL tube contains 1.4 ceramic spheres, 0.1 mm silica spheres, and one 4 mm glass bead.
- Key applications:
- Suitable for all sample types, Lysing matricies
- Suitable for all sample types, Lysing matricies
- Instrument Compatibility:
- FastPrep-24; FastPrep 120; FastPrep-96
- FastPrep-24; FastPrep 120; FastPrep-96
- Format:
- Matrix A: Zirconium sphere, Banded Satellite; Garnet Flakes, Block, Sharp angular grains. Matrix B, Spherical. Matrix C Spherical. Matrix D, Spherical. Matrix E, Spherical.
- Matrix A: Zirconium sphere, Banded Satellite; Garnet Flakes, Block, Sharp angular grains. Matrix B, Spherical. Matrix C Spherical. Matrix D, Spherical. Matrix E, Spherical.
- Adapter compatibility:
- QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)