To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Calibration:pH with calibration editing option, relative mV (RmV), ORP, and temperature
- Description:Orion™ Versa Star™ pH/mV/RmV/ORP/Temperature Measurement Module
- Operating temperature range:5…45 °C (41…113 °F)
- Temperature accuracy:±0.1°C
- Temperature resolution:0.1°C, 0.1°F
- Temperature range:–5 to 105°C (22 to 221°F)
- Input:BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), BNC (pH), 8-pin Mini-DIN (temperature), reference pin-tip
- IP rating:IP-54
- Alarm:Set point alarm, high/low limit alarm, calibration due alarm
- Calibration points:1 point relative mV (RmV) mode; 1 point ORP mode; 1 to 6 points pH mode
- Dimensions:12L x 3W x 9.5H cm (4.7 x 1.2 x 3.8")
- Weight:0.2 kg (0.5 lb.)
- mV accuracy:±0.2 mV or ±0.05% of reading, whichever is greater
- mV range:±2000.0 mV
- mV resolution:0.1 mV
- pH accuracy:±0.002 pH
- pH resolution:0.1, 0.01, 0.001 pH
- Cat. no.:89206-414
- Includes:Versa Star™ pH module and printed quick start guide
- Supplier No.:VSTAR-PH
Specifications
About this item
Add pH, mV, ORP, and temperature measurement capabilities to Versa Star™ meters using the Orion™ Versa Star™ pH Measurement Module.
- Measure pH, mV, relative mV, or ORP with temperature
- Perform up to a six point pH calibration using automatic buffer recognition of NIST, DIN, or user-defined buffer groups
- Calibration editing option allows individual points to be fixed without a full recalibration
- Customize measurements to show calibration history and pH electrode condition or simplify to show an extra large pH reading
- 10 methods per channel with password protection
- Three-year module warranty
Add pH, mV, ORP, and temperature measurement capabilities to an Orion™ Versa Star™ meter using the Orion™ Versa Star™ pH Measurement Module
Select from five Orion™ Versa Star™ measurement modules to customize the measurement capabilities of an Orion™ Versa Star™ meter. Measurement modules are interchangeable and can be added or removed from each Versa Star™ meter's four channels at any time.
Each Versa Star™ meter has four channels that can be customized with interchangeable measurement modules. The Versa Star™ pH module offers up to a six point pH calibration with easy to follow onscreen instructions, automatic buffer recognition of NIST, DIN, or user-defined buffer groups, and calibration editing option to fix individual points without a full recalibration.