To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Calibration:pH with calibration editing option, ion concentration (ISE) with calibration editing option, relative mV (RmV), ORP and temperature
- Description:Orion™ Versa Star™ pH/ISE/mV/RmV/ORP/Temperature Measurement Module
- ISE accuracy:±0.2 mV or ±0.05% of reading, whichever is greater
- Operating temperature range:5…45 °C (41…113 °F)
- Temperature accuracy:±0.1°C
- Temperature resolution:0.1°C, 0.1°F
- Temperature range:–5 to 105°C (22 to 221°F)
- Input:BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), BNC (pH or ISE), 8-pin Mini-DIN (temperature), reference pin-tip
- IP rating:IP-54
- Alarm:Set point alarm, high/low limit alarm, calibration due alarm
- Calibration points:1 point relative mV (RmV) mode; 1 point ORP mode; 1 to 6 points ISE mode; 1 to 6 points pH mode
- Dimensions:12L x 3W x 9.5H cm (4.7 x 1.2 x 3.8")
- ISE resolution:0.0001 minimum, 1 to 4 significant digits (user selectable)
- ISE range:0.0001 to 19990 with selectable units of ppm, Molar, mg/L, %, ppb, or none
- Weight:0.2 kg (0.5 lb.)
- mV accuracy:±0.2 mV or ±0.05% of reading, whichever is greater
- mV range:±2000.0 mV
- mV resolution:0.1 mV
- pH accuracy:±0.002 pH
- pH resolution:0.1, 0.01, 0.001 pH
- Cat. no.:89206-420
- Includes:Versa Star™ pH/ISE module and printed quick start guide
- Supplier No.:VSTAR-ISE
Specifications
About this item
Add pH, ion concentration, mV, ORP, and temperature measurement capabilities to a Versa Star™ meter using the Orion™ Versa Star™ pH/ISE Measurement Module.
- Measure pH, ion concentration, mV, relative mV, or ORP with temperature
- Advanced ISE features include non-linear selectable auto-blank, low range stability, and electrode ID with calibration verification
- 10 methods per channel with password protection
- Three-year module warranty
Add pH, ion concentration, mV, ORP, and temperature measurement capabilities to an Orion™ Versa Star™ meter using the Orion™ Versa Star™ pH/ISE Measurement Module
Perform up to a six point pH calibration using automatic buffer recognition of NIST, DIN, or user-defined buffer groups with choice to manually enter custom buffer values. Perform up to a six point ISE (Ion Selective Electrode) calibration with selectable units of ppm, M, mg/L, %, ppb, or none. Customize measurements to show calibration history and pH electrode condition or simplify to show an extra large pH or ion concentration reading.
Each Orion™ Versa Star™ meter has four channels that can be customized with interchangeable measurement modules. The Versa Star™ pH/ISE module offers up to six point pH and ISE calibrations and advanced ISE features, including incremental techniques (single and double known addition and subtraction, analate addition and subtraction), low range stability, and electrode ID with calibration verification.