Specifications
- Description:Orion™ Versa Star™ RDO/DO/Temperature Measurement Module
- Cat. no.:89206-418
- Includes:Versa Star™ RDO/DO Module and Printed Quick Start Guide
- Supplier No.:VSTAR-RD
Specifications
About this item
Add dissolved oxygen and temperature measurement capabilities to Versa Star™ meter using the Orion™ Versa Star™ Dissolved Oxygen/RDO Measurement Module.
- Calibrate dissolved oxygen using water-saturated air, air-saturated water, custom value (Winkler titration), and zero point options
- Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input, and built-in barometer for barometric pressure readings
- Customize measurements to show calibration history or simplify to show an extra large dissolved oxygen reading
- Module offers a three-year replacement warranty
Add dissolved oxygen and temperature measurement capabilities to Orion™ Versa Star™ meters using the Orion™ Versa Star™ Dissolved Oxygen/RDO Measurement Module. Each Versa Star™ meter has four channels that can be customized with interchangeable measurement modules. The Orion™ Versa Star™ RDO/DO module offers the flexibility to use Orion™ RDO optical or polarographic probes with automatic sensor recognition and calibration using water-saturated air, air-saturated water, custom value using Winkler titration, and zero point options. Measure dissolved oxygen in percent saturation or mg/L with temperature using Orion™ RDO optical DO probes or Orion polarographic DO probes and automatic sensor recognition.