About this item
Reliably measure pH, ion concentration, mV, ORP, and temperature with the Orion™ Star™ A214 pH/ISE Benchtop Meter for advanced laboratory analysis and interfacing.
- Calibration editing option allows individual points to be fixed without a full recalibration
- Advanced ISE features include non-linear selectable auto-blank and low range stability
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Capture critical readings with measurement stability indicator and selectable read modes
- Auto-Read mode holds a stable reading, Continuous mode shows changing readings with hold option, and Timed mode logs data at specific intervals
- Calibration log stores ten most recent calibrations
- Three-year meter warranty
Record accurate and reliable pH, ion concentration, mV, ORP, and temperature measurements with the Orion™ Star™ A214 pH/ISE Benchtop Meter, ideal for a wide range of applications and advanced pH or ion analysis in the laboratory.
These easy to use, dependable laboratory meters offer advanced features and functions. Selectable read modes and stability indicator provide flexibility and control over testing. All critical data is shown with measurements, including temperature, date and time, read type, measurement stability, active calibration information, and electrode condition icon. Onscreen calibration and setup instructions can be displayed in several local languages with new languages added through software updates. Navigation is fast and simple due to the comprehensive keypad with menu-specific function keys, dual purpose scroll/shortcut keys, plus power, measure, log view, and stirrer keys that ensure easy operation.
All setup menus are in one location with direct access using a single shortcut key to quickly access, evaluate, and change measurement and instrument settings. Perform up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values. Perform up to a five point ISE (Ion Selective Electrode) calibration with selectable units of ppm, M, mg/L, %, ppb, or none. Select from a wide variety of Orion ion selective electrodes, including electrodes that measure ammonia, ammonium, bromide, cadmium, calcium, carbon dioxide, chloride, chlorine, cupric, cyanide, fluoride, fluoroborate, iodide, lead, nitrate, potassium, silver, sodium, sulfate, surfactant, and thiocyanate concentrations. Eliminate the need for magnetic stir bars using the meter-controlled stirrer probe to uniformly mix solutions. Flexible power options with universal power adapter or optional AA batteries. IP54-rated housing resists dust and splashes, and can be wall-mounted. Non-volatile meter memory ensures data and settings are protected, even with loss of power.
Perform up to 5 point pH and ISE calibrations, log up to 2000 data point sets with time/date stamp, and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion™ Star Com software. Use the included electrode stand to easily place sensors in samples and simplify storage to minimize breakage.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 89206-336
- 89206-338
- 89206-332
- 89206-334
- 89206-330
- ISE accuracy:
- ±0.2 mV or ±0.05% of reading whichever is greater
- ±0.2 mV or ±0.05% of reading whichever is greater
- ±0.2 mV or ±0.05% of reading whichever is greater
- ±0.2 mV or ±0.05% of reading whichever is greater
- ±0.2 mV or ±0.05% of reading whichever is greater
- Temperature resolution:
- 0.1°C, 0.1°F
- 0.1°C, 0.1°F
- 0.1°C, 0.1°F
- 0.1°C, 0.1°F
- 0.1°C, 0.1°F
- Temperature accuracy:
- ±0.1°C
- ±0.1°C
- ±0.1°C
- ±0.1°C
- ±0.1°C
- Operating temperature range:
- 5…45 °C (41…113 °F)
- 5…45 °C (41…113 °F)
- 5…45 °C (41…113 °F)
- 5…45 °C (41…113 °F)
- 5…45 °C (41…113 °F)
- Memory:
- Non-volatile memory preserves data log, calibration log, and meter settings
- Non-volatile memory preserves data log, calibration log, and meter settings
- Non-volatile memory preserves data log, calibration log, and meter settings
- Non-volatile memory preserves data log, calibration log, and meter settings
- Non-volatile memory preserves data log, calibration log, and meter settings
- No. of channels:
- 1
- 1
- 1
- 1
- 1
- Calibration:
- pH with calibration editing option, ion concentration (ISE) with calibration editing option, relative mV (RmV), ORP, and temperature
- pH with calibration editing option, ion concentration (ISE) with calibration editing option, relative mV (RmV), ORP, and temperature
- pH with calibration editing option, ion concentration (ISE) with calibration editing option, relative mV (RmV), ORP, and temperature
- pH with calibration editing option, ion concentration (ISE) with calibration editing option, relative mV (RmV), ORP, and temperature
- pH with calibration editing option, ion concentration (ISE) with calibration editing option, relative mV (RmV), ORP, and temperature
Specifications
Related Information
S-STARA214-E-0811-RevA_WEB.pdf
Datasheet
VWR GHS Compliance Letter (US)